Iodacetamid
Lieferant: G Bioscienes
|
Achtung
|
Synonyme:
Iodessigsäureamid
786-228
786-228EA
132
EUR
786-228
Iodacetamid
Iodacetamid
Formel:
ICH₂CONH₂ Molecular Weight: 184.96 g/mol Siedepunkt: 266 Schmelzpunkt: 91…94 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00008028 CAS-Nummer: 144-48-9 EINECS: 205-630-1 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...