Pep-Extract™
Lieferant: G Bioscienes
Following digestion of proteins with proteases, the addition of Pep-Extract™ rapidly elutes digested peptides in under 20 minutes. The eluted peptides can be used directly in mass spectrometry analysis without any additional clean up. Contains sufficient for 500 protein spots.