Dry buffer packs, Carbonate-Bicarbonate Buffer, JAW™
Lieferant: G Bioscienes
786-291
786-291EA
208
EUR
786-291
Dry buffer packs, Carbonate-Bicarbonate Buffer, JAW™
Puffer
Biologische Puffer
Premixed buffer powders that simply require adding deionised water to generate a ready-to-use buffer.
JAW™ Carbonate-Bicarbonate Buffer packs each make one litre of: 0,2M carbonate-bicarbonate buffer (at a pH of 9,4).