Brij® 35
Lieferant: G Bioscienes
|
Achtung
|
Synonyme:
Brij® L23
786-521
786-351
786-521EA
139
EUR
786-521
786-351
Brij® 35
Brij® 35
Formel:
C₅₈H₁₁₈O₂₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00080891 CAS-Nummer: 9002-92-0 EINECS: 500-002-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...