CDI amine reactive resin
Lieferant: G Bioscienes
CDI amine reactive agarose consists of 6% cross-linked agarose activated with CDI (1,1’-carbonyl diimidazole) to form reactive imidazole carbamates.
- Proven coupling chemistry and stable linkages
- Easy to use
- Couple in inorganic buffers for insoluble molecules
- Couple proteins and peptides
- Couple primary amine containing ligands
Ideal for immobilising peptides, small organic molecules and certain proteins and reactions can occur in organic solvent making it suitable for water-insoluble ligands.