Hybridisation buffers, formamide-free, HYB-LINK™
Lieferant: G Bioscienes
786-405 786-406
786-405EA 335 EUR
786-405 786-406
Hybridisation buffers, formamide-free, HYB-LINK™
Puffer Elektrophorese-Puffer Hybridisierungspuffer
Hyb-LINK™ is a high performance formamide-free hybridisation buffer solution for Northern, Southern and Dot Blots, which increases sensitivity of random-primer DNA probes.
  • Reliable and accurate hybridisation
  • Increased sensitivity
  • Compatible with both RNA and DNA Probes