Hybridisation buffers, formamide-free, HYB-LINK™
Lieferant: G Bioscienes
786-405
786-406
786-405EA
335
EUR
786-405
786-406
Hybridisation buffers, formamide-free, HYB-LINK™
Puffer
Elektrophorese-Puffer
Hybridisierungspuffer
Hyb-LINK™ is a high performance formamide-free hybridisation buffer solution for Northern, Southern and Dot Blots, which increases sensitivity of random-primer DNA probes.
- Reliable and accurate hybridisation
- Increased sensitivity
- Compatible with both RNA and DNA Probes