Adenosylhomocysteine nucleosidase
Lieferant: G Bioscienes
786-427
786-427EA 717 EUR
786-427
Adenosylhomocysteine nucleosidase
Enzyme
This recombinant adenosylhomocysteine nucleosidase (10 µM) converts S-adenosylhomocysteine to S-(deoxy-D-ribos-5-yl)-L-homocysteine and adenine.