DATP [100MM] 1 * 25 mL
Lieferant: G Bioscienes
dATP aqueous solution at pH7,0 in 100mM concentration. For research use in PCR, long PCR, RT-PCR, cDNA synthesis, primer extension, DNA sequencing and DNA labeling. 1 * 25 mL