Coomassie® Brillantblau R 250 Gel-Elektrophorese-Färbung

Lieferant: G Bioscienes

Synonyme: Brillantblau R, Hydrogen[4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulfonatobenzyl)amino]benzhydrylen]cyclohexa-2,5-dien-1-yliden](ethyl)(m-sulfonatobenzyl)ammonium Mononatriumsalz, Coomassie Blau R 250, Xylenbrillantcyanin, Serva Blue R, Brillantindocyanin 6B, Acid Blue 83

786-495
786-495EA 106 EUR
786-495
Coomassie® Brillantblau R 250 Gel-Elektrophorese-Färbung
Coomassie® Brillantblau R 250

G-Biosciences' Coomassie Brilliant Blue (CBB) G-250 and R-250 Stain are based on a colloidal Coomassie stain. CBB G-250 and R-250 stain proteins with high band visibility. The staining of gels with CBB G-250 and R-250 allows the examination of protein bands even during the staining process.


After the staining process, the band intensity may be further enhanced by de-staining the stained gel in our CBB De-Staining Solution or 30% Methanol. CBB G-250 and R-250 Stains have a sensitivity of staining 8-10ng protein/band (e.g., 8-10ng of BSA is visible in 4-20% SDS acrylamide gels). The supplied volume is suitable for 40-50 mini gels (8 x 10cm) & 20-25 large gels (12 x 15cm).


G-Biosciences' Coomassie Brilliant Blue R-250 and G-250 are both offered in dry powder form and convenient ready-to-use solutions.
A destaining solution is also offered.

Formel: C₄₅H₄₄N₃NaO₇S₂
Molecular Weight: 825.98 g/mol
Schmelzpunkt: 174…180 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041762
CAS-Nummer: 6104-59-2
EINECS: 228-060-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR