Coomassie Brilliant Blue Destaining Solution, G-Biosciences
Lieferant: G Bioscienes
786-499
786-499EA
143
EUR
786-499
Coomassie Brilliant Blue Destaining Solution, G-Biosciences
Elektrophorese-Reagenzien
Reagenzien zur Entfernung von Farbstoffen
After Coomassie Brilliant Blue staining process, the band intensity may be further enhanced by de-staining the stained gel in our CBB De-Staining Solution.
- Contains Methanol and Acetic acid