Tween® 20 (Polysorbat)

Lieferant: G Bioscienes

786-517 786-518
786-517EA 111 EUR
786-517 786-518
Tween® 20 (Polysorbat)
Tween® 20 (Polysorbat)

Tween® 20 is commonly used non ionic detergent for solubilising membrane proteins during isolation of membrane-protein complexes.

Formel: C₅₈H₁₁₄O₂₆
MDL: MFCD00165986
CAS-Nummer: 9005-64-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60d52c1043214149edc1c697a3ffffe58f6c025f&vid=0633c24839d746937941c948790c95906898733e&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR