Destain solutions
Lieferant: G Bioscienes
786-526
786-527
786-526EA
137
EUR
786-526
786-527
Destain solutions
Elektrophorese-Reagenzien
Reagenzien zur Entfernung von Farbstoffen
G-Biosciences' Destain I contains approximately 45% methanol and 9% glacial acetic acid. G-Biosciences' Destain II contains approximately 5% methanol and 7% acetic acid.