Blue Dextran Ladepuffer 

Lieferant: G Bioscienes

Synonyme: Dextran, blue

786-556 786-557 786-574
786-556EA 66.1 EUR
786-556 786-557 786-574
Blue Dextran Ladepuffer 
Blue Dextran

A proprietary mixture of Formaldehyde, EDTA and Blue Dextran that is designed for use in loading DNA samples onto denaturing polyacrylamide gels.


Commonly used in loading DNA samples onto denaturing polyacrylamide gels.

Formel: (C₂₉H₂₁N₇O₁₂S₃·3Na)n
MDL: MFCD00130661
CAS-Nummer: 87915-38-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR