Immobilised avidin for biotin purification
Lieferant: G Bioscienes
786-594
786-593
786-594EA
1690
EUR
786-594
786-593
Immobilised avidin for biotin purification
Protein-Aufbereitungs
Protein Purification Resins
This avidin resin is designed for the single step small and large scale affinity purification of proteins and antibodies with a biotin tag. The resin can also be used for immunoprecipitations using biotin labelled antibodies.