Subcellular Protein Fractionation Kit for Cultured Cells
Lieferant: Thermo Fisher Scientific
78840
78840.EA 642 EUR
78840.
Subcellular Protein Fractionation Kit for Cultured Cells
Puffer Extraktionspuffer
This kit is ideal for segregation and enrichment of proteins from five different cellular compartments.
  • Concise - extract functional cytoplasmic, membrane, nuclear soluble, chromatin-bound and cytoskeletal protein fractions in less than 3 hours
  • Convenient - perform a simple procedure without using ultracentrifugation over gradients
  • Complete - obtain protein fractions from a single kit
  • Cultured cells - developed and optimized for use with cultured mammalian cells
  • Compatible - use extracts for downstream applications such as protein assays, Western blotting, protein assays, electrophoretic mobility shift assays, reporter-gene and enzyme-activity assays
Lieferumfang: Kit includes four extraction buffers, a stabilised nuclease and halt protease inhibitor cocktail.