Organic reference standard, 4-Hydroxy-4-methyl-2-pentanone, analytical standard, Supelco®
Lieferant: Merck
78891-5ML
78891-5MLEA
226
EUR
78891-5ML
Organic reference standard, 4-Hydroxy-4-methyl-2-pentanone, analytical standard, Supelco®
Standards
Organische Standards
4-Hydroxy-4-methyl-2-pentanone may be used as an internal standard in the analysis of volatile odor-active compounds present in wine using solid phase extraction with gas chromatography (SPE-GC).