Supelco® TraceCERT® Organic Reference Standard, Amino Acids Mix Solution
Lieferant: Merck
79248-5X2ML
79248-5X2MLEA
239
EUR
79248-5X2ML
Supelco® TraceCERT® Organic Reference Standard, Amino Acids Mix Solution
Standards
Organische Standards
- GHS signal: GHS05
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Bitte beachten P234 - P390