Cesium standard for ICP, TraceCERT®, Supelco®
Lieferant: Merck
79261-100ML
79261-100MLEA
156
EUR
79261-100ML
Cesium standard for ICP, TraceCERT®, Supelco®
Standards
Kationischer Standards
Cäsium-Standards
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034