DAABD-AE ≥95,0% (durch HPLC), LiChropur™ Derivatisierungsreagenz, für die HPLC, Supelco®

Lieferant: Merck

Synonyme: 4-[2-(N,N-Dimethylamino)ethylaminosulphonyl]-7-(2-aminoethylamino)-2,1,3-benzoxadiazole

79291-100MG
79291-100MGEA 535 EUR
79291-100MG
DAABD-AE ≥95,0% (durch HPLC), LiChropur™ Derivatisierungsreagenz, für die HPLC, Supelco®
DAABD-AE

4-[2-(N,N-dimethylamino)ethylaminosulfonyl]-7-(2-aminoethylamino)-2,1,3-benzoxadiazole (DAABD-AE) is a new derivatization reagent for rapid and specific liquid chromatography tandem mass spectrometric quantification of pristanic acid, phytanic acid, and very long chain fatty acids (i.e., hexacosanoic, tetracosanoic, and docosanoic acids) in plasma.


Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.

Formel: C₁₂H₂₀N₆O₃S
Molecular Weight: 328,39 g/mol
CAS-Nummer: 913253-56-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1ee49ed1d5406cf195a5bec1c2021f06d39d6f4&vid=c6b8c20b79f025d64394653f0ba43813bf50b67a&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR