Amino-functionalized silica gel, sphärisch 40-75 μm particle size, Supelco®
Lieferant: Merck
|
Achtung
|
79297-100G
79297-1KG
79297-100GEA
203
EUR
79297-100G
79297-1KG
Amino-functionalized silica gel, sphärisch 40-75 μm particle size, Supelco®
Amino-functionalized silica gel
Lagertemperatur:
Raumtemperatur |
CAS-Nummer:
126850-14-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...