Supelco® TraceCERT® Organic Reference Standard, Ethylparaben
Lieferant: Merck
79577-50MG
79577-50MGEA
146
EUR
79577-50MG
Supelco® TraceCERT® Organic Reference Standard, Ethylparaben
Standards
Organische Standards
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.