Organic reference standard, Phloridzin dihydrate, analytical standard, Supelco®
Lieferant: Merck
79589-50MG
79589-50MGEA 65.2 EUR
79589-50MG
Organic reference standard, Phloridzin dihydrate, analytical standard, Supelco®
Standards Organische Standards
This standard is used to induce experimental glycosuria.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=011b1dbb9ed007bf23f5b373bfa1f56b8943907c&vid=011b1dbb9ed007bf23f5b373bfa1f56b8943907c&prid=web&clid=SAPDAM