Dimethyltrisulfid, Supelco®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

79592-10MG-F
79592-10MG-FEA 79.4 EUR
79592-10MG-F
Dimethyltrisulfid, Supelco®
Dimethyltrisulfid
Formel: C₂H₆S₃
MDL: MFCD00039808
CAS-Nummer: 3658-80-8
EINECS: 222-910-9
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55069bf33a29b9676ccf91cd34433363dc5fab30&vid=32e3fbd5ab2474d3c85949ca344d7055fdb36e07&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR