Sulfate standard for IC, TraceCERT®, Supelco®
Lieferant: Merck
79735-100ML
79735-100MLEA
86.1
EUR
79735-100ML
Sulfate standard for IC, TraceCERT®, Supelco®
Standards
Anionische Standards
Sulfat-Standards
Standards: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.