Organic reference standard, Isoborneol, analytical standard, Supelco®
Lieferant: Merck
79856-100MG
79856-100MGEA
48.3
EUR
79856-100MG
Organic reference standard, Isoborneol, analytical standard, Supelco®
Standards
Organische Standards
Isoborneol is a monoterpene and a known antioxidant, which is a component of several plant essential oils.