2-Aminophenol zur Synthese, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ200

8.00419.9010
8.00419.9010EA 543 EUR
8.00419.9010
2-Aminophenol zur Synthese, Sigma-Aldrich®
2-Aminophenol
Formel: C₆H₇NO
MDL: MFCD00007690
CAS-Nummer: 95-55-6
UN: 2512
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Solubility 17 g/l (20 °C)
Melting Point 172 - 174 °C (sublimed)
Molar Mass 109.12 g/mol
Bulk Density 600 kg/m3
Boiling Point 150 °C (15 hPa)
Vapor Pressure 14 hPa (153 °C)
Flash Point >175 °C
Density 1.328 g/cm3 (20 °C)
pH 6.5 - 7.5 (10 g/l, H2O, 20 °C)
Ignition Point 190 °C
Synonyms 2-Hydroxy aniline, 2-Amino-1-hydroxy benzene
Hill Formula C6H7NO
Chemical Formula 2-(NH2)C6H4OH


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR