3-Nitrobenzoesäure zur Synthese, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ200

8.00645.9025
8.00645.9025EA 0 EUR
8.00645.9025
3-Nitrobenzoesäure zur Synthese, Sigma-Aldrich®
3-Nitrobenzoesäure
Formel: C₇H₅NO₄
MDL: MFCD00007251
CAS-Nummer: 121-92-6

Bestellen

Spezifikation Testergebnisse

Solubility 3 g/l (25 °C)
Melting Point 139 - 142 °C
Molar Mass 167.12 g/mol
Bulk Density 700 kg/m3
Flash Point 190 °C
Density 1.49 g/cm3 (20 °C)
pH 3 (5 g/l, H2O, 20 °C) (aqueous suspension)
Ignition Point 490 °C
Hill Formula C7H5NO4
Chemical Formula 3-(NO2)C6H4COOH


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR