Diethylethylmalonat, Sigma-Aldrich®

Lieferant: Merck


MQ200

8.00906.0250
8.00906.0250EA 59.1 EUR
8.00906.0250
Diethylethylmalonat, Sigma-Aldrich®
Diethylethylmalonat
Formel: C₉H₁₆O₄
Molecular Weight: 188.22 g/mol
MDL: MFCD00009167
CAS-Nummer: 133-13-1
EINECS: 205-093-3

Bestellen

Spezifikation Testergebnisse

Solubility 2.87 g/l (20 °C)
Molar Mass 188.22 g/mol
Boiling Point 204 - 206 °C (1013 hPa)
Flash Point 88 °C
Refractive Index 1.4158 (20 °C, 589 nm)
Density 1.00 g/cm3 (20 °C)
Synonyms Ethylmalonic acid diethyl ester
Hill Formula C9H16O4
Chemical Formula C2H5CH(COOC2H5)2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR