2-Amino-2-thiazolin

Lieferant: Merck

Synonyme: 4,5-Dihydrothiazol-2-amine

M-Clarity™ Qualitätsstufe = MQ200

8.01313.0050
8.01313.0050EA 305 EUR
8.01313.0050
2-Amino-2-thiazolin
2-Amino-2-thiazolin
Formel: C₃H₆N₂S
Molecular Weight: 102.16 g/mol
MDL: MFCD00005313
CAS-Nummer: 1779-81-3
EINECS: 217-224-1

Bestellen

Spezifikation Testergebnisse

Solubility (20 °C) soluble
Melting Point 76 - 79 °C
Molar Mass 102.16 g/mol
Hill Formula C3H6N2S


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR