2-(Biphenyl-4-yl)-2-propanol

Lieferant: Merck

8.01965.0010
8.01965.0010EA 0 EUR
8.01965.0010
2-(Biphenyl-4-yl)-2-propanol
2-(Biphenyl-4-yl)-2-propanol
Formel: C₁₅H₁₆O
MDL: MFCD00014390
CAS-Nummer: 34352-74-4
EINECS: 251-955-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR