Biphenyl-2,2'-dicarbonsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
[1,1'-Biphenyl]-2,2'-dicarboxylic acid, Diphenic acid, 2,2'-Biphenyldicarboxylic acid
M-Clarity™ Qualitätsstufe = MQ200
8.02256.0050
8.02256.0050EA
114
EUR
8.02256.0050
Biphenyl-2,2'-dicarbonsäure, Sigma-Aldrich®
Biphenyl-2,2'-dicarbonsäure
Formel:
C₁₄H₁₀O₄ Molecular Weight: 242,23 g/mol |
MDL:
MFCD00002464 CAS-Nummer: 482-05-3 EINECS: 207-576-4 |
Spezifikation Testergebnisse
Solubility | (20 °C) insoluble |
Melting Point | 225 - 228 °C |
Molar Mass | 242.23 g/mol |
Bulk Density | 470 kg/m3 |
pH | 4 - 5 (H2O, 20 °C) |
Ignition Point | 390 °C |
Synonyms | Diphenic acid, Diphenyl-2,2-dicarboxylic acid |
Hill Formula | C14H10O4 |
Chemical Formula | 2-(HOOC)C6H4C6H4-2-(COOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...