4-Chlorbutyrylchlorid

Lieferant: Merck

Synonyme: 4-Chlorobutanoyl Chloride

8.02509.1000
8.02509.1000EA 0 EUR
8.02509.1000
4-Chlorbutyrylchlorid
4-Chlorbutyrylchlorid
Formel: C₄H₆Cl₂O
Molecular Weight: 141 g/mol
MDL: MFCD00000754
CAS-Nummer: 4635-59-0
EINECS: 225-059-1
UN: 2927
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR