(±)-α-Chlorphenylacetylchlorid

Lieferant: Merck

8.02699.0050
8.02699.0050EA 0 EUR
8.02699.0050
(±)-α-Chlorphenylacetylchlorid
(±)-α-Chlorphenylacetylchlorid
Formel: C₈H₆Cl₂O
Molecular Weight: 189,04 g/mol
CAS-Nummer: 2912-62-1
EINECS: 220-826-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR