Cystaminiumdichlorid, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Cystaminium dichloride, 2,2′-Diaminodiethyl disulphide dihydrochloride, 2,2'-Disulphanediyldiethanamine dihydrochloride, Bis(2-aminoethyl) disulphide dihydrochloride, Decarboxycystine dihydrochloride, Cystamine (dihydrochloride), {2-[(2-Aminoethyl)disulphanyl]ethyl}amine dihydrochloride, 2,2′-Dithiobis(ethylamine) dihydrochloride

M-Clarity™ Qualitätsstufe = MQ200

8.02834.0025 8.02834.0100
8.02834.0025EA 59.7 EUR
8.02834.0025 8.02834.0100
Cystaminiumdichlorid, Sigma-Aldrich®
Cystaminiumdichlorid
Formel: C₄H₁₂N₂S₂·2HCl
Lagertemperatur: Raumtemperatur
MDL: MFCD00012905
CAS-Nummer: 56-17-7
EINECS: 200-260-7

Bestellen

Spezifikation Testergebnisse

Solubility (20 °C) soluble
Melting Point 218 - 221 °C
Molar Mass 225.19 g/mol
Synonyms Cystamine dihydrochloride
Hill Formula C4H14Cl2N2S2
Chemical Formula [H2NCH2CH2SSCH2CH2NH2] * 2 HCl


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR