1,8-Diazabicyclo[5.4.0]-7-undecen ≥98%

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ200

8.03282.0025 8.03282.2500 8.03282.0100 8.03282.9050
8.03282.0025EA 42.2 EUR
8.03282.0025 8.03282.2500 MERC8.03282.0100 8.03282.9050
1,8-Diazabicyclo[5.4.0]-7-undecen ≥98%
1,8-Diazabicyclo[5.4.0]-7-undecen
Formel: C₉H₁₆N₂
MDL: MFCD00006930
CAS-Nummer: 6674-22-2
UN: 2922
ADR: 8 (6.1),II

Bestellen

Spezifikation Testergebnisse

Solubility Soluble at 20 °C
Molar Mass 152.24 g/mol
Boiling Point 261 °C (1013 hPa)
Vapor Pressure 0.02 hPa (25 °C)
Flash Point 116 °C
Explosion Limit 1.1 - 6.5 %(V)
Density 1.02 g/cm3 (20 °C)
pH 12.8 (10 g/l, H2O, 20 °C)
Ignition Point 260 °C
Ignition temperature 260 °C
pH value 12,8
Vapor pressure 0,02 hPa
Storage temperature +15...+25 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR