α-Jonon, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(E)-4-(2,6,6-Trimethylcyclohex-2-en-1-yl)but-3-en-2-one
M-Clarity™ Qualitätsstufe = MQ200
8.04778.0050
8.04778.0050EA
38.4
EUR
8.04778.0050
α-Jonon, Sigma-Aldrich®
α-Jonon
Formel:
C₁₃H₂₀O Molecular Weight: 192,3 g/mol |
MDL:
MFCD00001565 CAS-Nummer: 127-41-3 EINECS: 204-841-6 |
Spezifikation Testergebnisse
Solubility | 110 mg/l (20 °C) |
Molar Mass | 192.3 g/mol |
Boiling Point | 237 °C (1013 hPa) |
Vapor Pressure | 0.0013 hPa (20 °C) |
Flash Point | 117 °C |
Refractive Index | 1.5 (20 °C, 589 nm) |
Density | 0.93 g/cm3 (20 °C) |
Hill Formula | C13H20O |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...