N-Methyl-2-pyrrolidon (NMP), EMPLURA®, Supelco®

Lieferant: Merck
Gefahr

Synonyme: 1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, 1-Methyl-2-pyrrolidone, NMP, Methyl Pyrrolidone

M-Clarity™ Qualitätsstufe = MQ200

8.06072.1000 8.06072.2500 8.06072.9025 8.06072.9210 8.06072.9011 8.06072.9180
8.06072.1000EA 74.5 EUR
8.06072.1000 8.06072.2500 8.06072.9025 8.06072.9210 8.06072.9011 8.06072.9180
N-Methyl-2-pyrrolidon (NMP), EMPLURA®, Supelco®
N-Methyl-2-pyrrolidon (NMP)

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA® we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA® solvents provide adequate specifications with the most important parameters.

Formel: C₅H₉NO
Molecular Weight: 99.13 g/mol
Siedepunkt: 202
Schmelzpunkt: –24 °C
Dichte: 1.033 g/cm³ (20 °C)
Flammpunkt: 91
Lagertemperatur: Raumtemperatur
MDL: MFCD00003193
CAS-Nummer: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR