N-Methylformanilid, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ200
8.06169.0250
8.06169.0250EA
76.6
EUR
8.06169.0250
N-Methylformanilid, Sigma-Aldrich®
N-Methylformanilid
Formel:
C₈H₉NO Molecular Weight: 135,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003283 CAS-Nummer: 93-61-8 EINECS: 202-262-3 |
Spezifikation Testergebnisse
Hill Formula | C₈H₉NO |
Chemical formula | HCON(CH₃)C₆H₅ |
Molar Mass | 135.17 g/mol |
Boiling point | 243 - 244 °C (1013 hPa) |
Density | 1.095 g/cm³ (20 °C) |
Flash point | 126 °C |
Melting Point | 8 - 13 °C |
pH value | 3.5 - 5.5 (H₂O, 20 °C) (saturated aqueous solution) |
Vapor pressure | 2.66 Pa (25 °C) |
Refractive Index | 1.5593 (20 °C, 589 nm) |
Solubility | 10.3 g/l (25 °C) |
Storage class | 10 - 13 Other liquids and solids |
WGK | WGK 3 highly water endangering |
R Phrase | R 38 |
S Phrase | S 25 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...