Nickel(II)chlorid zur Synthese, Sigma-Aldrich®

Lieferant: Merck

8.06722.0100
8.06722.0100EA 102 EUR
8.06722.0100
Nickel(II)chlorid zur Synthese, Sigma-Aldrich®
Nickel(II)chlorid

Bestellen

Spezifikation Testergebnisse

Solubility (20 °C) soluble
Melting Point 1009 °C
Molar Mass 129.6 g/mol
Vapor Pressure 1.33 hPa (671 °C)
Density 3.55 g/cm3
pH 4 (500 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR