Pyrrol zur Synthese, Sigma-Aldrich®
Lieferant: Merck
8.07492.0025
8.07492.0100
8.07492.0010
8.07492.0025EA
30.1
EUR
8.07492.0025
8.07492.0100
8.07492.0010
Pyrrol zur Synthese, Sigma-Aldrich®
Spezifikation Testergebnisse
Solubility | 60 g/l (20 °C) |
Melting Point | -24 °C |
Molar Mass | 67.09 g/mol |
Boiling Point | 130 - 131 °C (1013 hPa) |
Vapor Pressure | 8.7 hPa (20 °C) |
Flash Point | 39 °C |
Refractive Index | 1.5082 (20 °C, 589 nm) |
Explosion Limit | 3.10 - 14.8 %(V) |
Density | 0.97 g/cm3 (20 °C) |
pH |
|
Ignition Point | 550 °C |
Hazard Class | 3 |
Hazard Class Description | Flammable liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...