1-Octadecanol zur Synthese
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
8.07680.0100
8.07680.1000
8.07680.0100EA
31.3
EUR
8.07680.0100
8.07680.1000
1-Octadecanol zur Synthese
1-Octadecanol
Formel:
H₃C(CH₂)₁₆CH₂OH Siedepunkt: 336 °C (1013 hPa) Schmelzpunkt: 56…57 °C Dichte: 0,805…0.815 g/cm³ (60 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002823 CAS-Nummer: 112-92-5 EINECS: 204-017-6 Merck Index: 12,08960 |
Spezifikation Testergebnisse
Hill Formula | C₁₈H₃₈O |
Molar Mass | 270.49 g/mol |
Boiling point | 330 - 360 °C (1013 hPa) |
Density | 0.805 - 0.815 g/cm³ (60 °C) |
Flash point | 195 °C |
Ignition temperature | 230 °C DIN 51794 |
Melting Point | 55 - 60 °C |
Vapor pressure |
|
Bulk density | 300 kg/m³ |
Solubility | (20 °C) practically insoluble |
LD 50 oral |
LD50 Rat |
Storage class | 10 - 13 Other liquids and solids |
WGK | NWG not water endangering |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...