Titanyl(IV)acetylacetonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Titan(IV)oxidacetylacetonat, Oxobis(2,4-pentandionato-O,O')titan(IV), Titan(IV)oxyacetylacetonat, Titan(IV)oxidbis(2,4-pentandionat)

M-Clarity™ Qualitätsstufe = MQ200

8.08305.0025 8.08305.0100
8.08305.0025EA 153 EUR
8.08305.0025 8.08305.0100
Titanyl(IV)acetylacetonat, Sigma-Aldrich®
Titanyl(IV)acetylacetonat
Formel: C₁₀H₁₄O₅Ti
Molecular Weight: 262.1 g/mol
MDL: MFCD00013505
CAS-Nummer: 14024-64-7
EINECS: 237-861-9

Bestellen

Spezifikation Testergebnisse

Solubility (20 °C) practically insoluble
Melting Point 200 °C
Molar Mass 262.08 g/mol
Bulk Density 500 kg/m3
Boiling Point 159 - 160 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR