1-Chlorethylethylcarbonat zur Synthese
Lieferant: Merck
Synonyme:
Carbonic acid 1-Chloroethyl ethyl ester, 1-Chloroethyl ethyl carbonate, 1-Chlorodiethyl Carbonate
8.10847.0100
8.10847.0100EA
0
EUR
8.10847.0100
1-Chlorethylethylcarbonat zur Synthese
1-Chlorethylethylcarbonat
Formel:
C₅H₉ClO₃ Molecular Weight: 152,58 g/mol |
MDL:
MFCD00040523 CAS-Nummer: 50893-36-2 EINECS: 256-832-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...