1,10-Didecyl-1,10-diaza-18-krone-6 (7,16-Didecyl-1,4,10,13-tetraoxa-7,16-diazacyclooctadecan, Kryptofix® 22 DD) zur Synthese, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

8.11963.0001
8.11963.0001EA 513 EUR
8.11963.0001
1,10-Didecyl-1,10-diaza-18-krone-6 (7,16-Didecyl-1,4,10,13-tetraoxa-7,16-diazacyclooctadecan, Kryptofix® 22 DD) zur Synthese, Sigma-Aldrich®
1,10-Didecyl-1,10-diaza-18-krone-6 (7,16-Didecyl-1,4,10,13-tetraoxa-7,16-diazacyclooctadecan, Kryptofix® 22 DD)
Formel: C₃₂H₆₆N₂O₄
Molecular Weight: 542,89 g/mol
CAS-Nummer: 79495-97-9
EINECS: 279-168-4

Bestellen

Spezifikation Testergebnisse

Solubility (20 °C) insoluble
Melting Point 36 - 39 °C
Molar Mass 542.88 g/mol
Synonyms 4,13-Didecyl-1,7,10,16-tetraoxa-4,13-diazacyclooctadecane
Hill Formula C32H66N2O4

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR