N-Phenylaza-15-krone-5 zur Synthese, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
13-Phenyl-1,4,7,10-tetraoxa-13-azacyclopentadecane, N-Phenyl-13-aza-1,4,7,10-tetraoxacyclopentadecane, N-Phenylaza-15-crown 5-Ether
M-Clarity™ Qualitätsstufe = MQ200
8.13885.0005
8.13885.0005EA
219
EUR
8.13885.0005
N-Phenylaza-15-krone-5 zur Synthese, Sigma-Aldrich®
N-Phenylaza-15-krone-5
Formel:
C₁₆H₂₅NO₄ Molecular Weight: 295,37 g/mol |
MDL:
MFCD00040513 CAS-Nummer: 66750-10-5 EINECS: 266-470-6 |
Spezifikation Testergebnisse
Solubility | (20 °C) slightly soluble |
Melting Point | 44 - 46 °C |
Molar Mass | 295.38 g/mol |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...