(2S,3S)-(+)-Diethyl-2,3-O-benzylidentartrat zur Synthese

Lieferant: Merck

8.14041.0005
8.14041.0005EA 0 EUR
8.14041.0005
(2S,3S)-(+)-Diethyl-2,3-O-benzylidentartrat zur Synthese
(2S,3S)-(+)-Diethyl-2,3-O-benzylidentartrat
Formel: C₁₅H₁₈O₆
CAS-Nummer: 141042-56-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR