Carbonylbis(triphenylphosphine)rhodium(I) chloride
Lieferant: Merck
8.14069.0001
8.14069.0001EA
0
EUR
8.14069.0001
Carbonylbis(triphenylphosphine)rhodium(I) chloride
Carbonylbis(triphenylphosphine)rhodium(I) chloride
Formel:
C₃₇H₃₀ClOP₂Rh |
MDL:
MFCD00044273 CAS-Nummer: 13938-94-8 |
Spezifikation Testergebnisse
Solubility | (20 °C) insoluble |
Melting Point | 224 - 227 °C decomposes |
Molar Mass | 690.95 g/mol |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...