2-Amino-2-methylpropanol Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (2-Hydroxy-1,1-dimethylethyl)ammoniumchlorid, 2-Amino-2-methyl-1-propanol Hydrochlorid, Isobutanolamin Hydrochlorid

M-Clarity™ Qualitätsstufe = MQ200

8.14099.0100
8.14099.0100EA 190 EUR
8.14099.0100
2-Amino-2-methylpropanol Hydrochlorid, Sigma-Aldrich®
2-Amino-2-methylpropanol Hydrochlorid
Formel: C₄H₁₂ClNO
Molecular Weight: 125,6 g/mol
MDL: MFCD00039086
CAS-Nummer: 3207-12-3
EINECS: 221-713-5

Bestellen

Spezifikation Testergebnisse

Solubility (20 °C) soluble
Melting Point 203 - 206 °C
Molar Mass 125.60 g/mol
Synonyms AMP hydrochloride
Hill Formula C4H12ClNO
Chemical Formula (CH3)2C(NH2)CH2OH * HCl

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR