2-(2-Nitroethyl)-1,3-dioxolan zur Synthese

Lieferant: Merck

Synonyme: 2-(2-Nitroethyl)[1,3]dioxolane

M-Clarity™ Qualitätsstufe = MQ200

8.14136.0001
8.14136.0001EA 0 EUR
8.14136.0001
2-(2-Nitroethyl)-1,3-dioxolan zur Synthese
2-(2-Nitroethyl)-1,3-dioxolan
Formel: C₅H₉NO₄
Molecular Weight: 147,13 g/mol
MDL: MFCD00040621
CAS-Nummer: 82891-99-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR