Polyethylenglycoldimethylether, M.W. 2,000 zur Synthese, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Poly(ethylene glycol) dimethyl ether Mn 250

M-Clarity™ Qualitätsstufe = MQ200

8.14172.0500
8.14172.0500EA 91.2 EUR
8.14172.0500
Polyethylenglycoldimethylether, M.W. 2,000 zur Synthese, Sigma-Aldrich®
Polyethylenglycoldimethylether
Formel: H₃C(OCH₂CH₂)nOCH₃
MDL: MFCD00147972
CAS-Nummer: 24991-55-7

Bestellen

Spezifikation Testergebnisse

Solubility (20 °C) soluble
Melting Point 49 - 53 °C
Molar Mass 2000 g/mol
Boiling Point 250 °C (1013 hPa)
Vapor Pressure  - 0.01 hPa (20 °C)
Flash Point 254 °C
Refractive Index 1.455 (60 °C, 589 nm)
Density 1.08 g/cm3 (60 °C)
pH (100 g/l, H2O, 20 °C) neutral
Ignition Point 385 °C DIN 51794
Viscosity kinematic 30 mm2/s (100 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR